missing translation for 'onlineSavingsMsg'
Learn More

8-Bromo-1-octene, 97%, Thermo Scientific Chemicals

Product Code. 15442209
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

8-Bromo-1-octene has been used in preparation of polymerizable ligand, required for the synthesis of quantum dot-labeled polymer beads. Grignard reagent derived from 8-bromo-1-octene has been used in the synthesis of (2S,3S,5R)-5-[(1R)-1-hydroxy-9-decenyl]-2-pentyltetrahydro-3-furanol. It has been used as building block in natural product synthesis. Also used for the synthesis of more complex pharmaceutical compounds, such as Dithienogermole (DTG) derivatives with varying alkyl chain lengths and pendant functionalities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Bromo-1-octene has been used in preparation of polymerizable ligand, required for the synthesis of quantum dot-labeled polymer beads. Grignard reagent derived from 8-bromo-1-octene has been used in the synthesis of (2S,3S,5R)-5-[(1R)-1-hydroxy-9-decenyl]-2-pentyltetrahydro-3-furanol. It has been used as building block in natural product synthesis. Also used for the synthesis of more complex pharmaceutical compounds, such as Dithienogermole (DTG) derivatives with varying alkyl chain lengths and pendant functionalities.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2695-48-9
Molecular Formula C8H15Br
Molecular Weight (g/mol) 191.112
MDL Number MFCD00000275
InChI Key SNMOMUYLFLGQQS-UHFFFAOYSA-N
Synonym 8-bromo-1-octene, 1-octene, 8-bromo, 7-octenyl bromide, 8-bromooctene, 8-azaquanine, bromo-8 octene-1, 8-bromo-oct-1-ene, acmc-1cmcf, oct-7-en-1-yl bromide
PubChem CID 75907
IUPAC Name 8-bromooct-1-ene
SMILES C=CCCCCCCBr

Specifications

Density 1.139 g/mL
Boiling Point 92°C (24 mmHg)
Flash Point 78°C (172°F)
Linear Formula Br(CH2)6CH=CH2
Refractive Index 1.467
Quantity 1 g
Beilstein 2234481
Formula Weight 191.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 8-Bromo-1-octene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.