missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ 700 Series Cemented Needle Syringes

This series of syringes is ideal for dispensing volumes from 0.5 up to 500μL.

Brand:  Hamilton™ DS80865

 View more versions of this product

Product Code. 11528034

  • 2010.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: a serialized syringe of your choice and a certificate of calibration.
This item is not returnable. View return policy

Description

Description

  • The original hand-fitted Hamilton™ syringe
  • Stainless steel plunger is manufactured to fit the glass barrel with a tolerance less than three thousandths of a millimeter
  • Unsurpassed syringe life
Specifications

Specifications

Not Autoclavable
22 G
0.72 mm
Microliter
50°C
10°C
500 μL
Borosilicate Glass, Stainless Steel
0.41 mm
2 in.
1000 psig (68.9 bar)
3
1/Pk.
Cemented
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.