missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 7-Hexadecyn-1-ol, 95%

Chemical Identifiers

CAS 822-21-9
Molecular Formula C16H30O
Molecular Weight (g/mol) 238.415
MDL Number MFCD00041709
InChI Key UFLJPANUONDNPT-UHFFFAOYSA-N
Synonym 7-hexadecyn-1-ol, 7-hexadecyne-1-ol, 7-hexadecyn-1-ol,
PubChem CID 549047
IUPAC Name hexadec-7-yn-1-ol
SMILES CCCCCCCCC#CCCCCCCO
View More Specs

Description

Description

Applications
7-Hexadecyn-1-ol is used in the preparation of 15-hexadecynoic acid. It acts as a reagent for pharmaceutical research and in chemical synthesis.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

822-21-9
238.415
UFLJPANUONDNPT-UHFFFAOYSA-N
549047
CCCCCCCCC#CCCCCCCO
C16H30O
MFCD00041709
7-hexadecyn-1-ol, 7-hexadecyne-1-ol, 7-hexadecyn-1-ol,
hexadec-7-yn-1-ol

Specifications

822-21-9
CH3(CH2)7C≡C(CH2)6OH
7-hexadecyn-1-ol, 7-hexadecyne-1-ol, 7-hexadecyn-1-ol,
CCCCCCCCC#CCCCCCCO
238.415
238.41
7-Hexadecyn-1-ol
C16H30O
MFCD00041709
UFLJPANUONDNPT-UHFFFAOYSA-N
hexadec-7-yn-1-ol
549047
95%
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.