missing translation for 'onlineSavingsMsg'
Learn More

7-Bromoheptanenitrile, 98%, Thermo Scientific Chemicals

Product Code. 11341817
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20965-27-9
Molecular Formula C7H12BrN
Molecular Weight (g/mol) 190.08
MDL Number MFCD00001982
InChI Key HVVQSKCGHAPHMV-UHFFFAOYSA-N
Synonym 7-bromoheptanonitrile, heptanenitrile, 7-bromo, 6-cyanohexyl bromide, 1-bromo-6-cyanohexane, 6-bromohexylcyanide, 6-bromohexyl cyanide, 7-bromo-heptanonitrile, 7-bromoheptan enitrile, 7-bromanylheptanenitrile, acmc-1cc07
PubChem CID 88737
IUPAC Name 7-bromoheptanenitrile
SMILES BrCCCCCCC#N

Specifications

Density 1.266
Boiling Point 114°C to 115°C (4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.475
Quantity 5 g
UN Number UN3276
Beilstein 1748070
Formula Weight 190.09
Percent Purity 98%
Chemical Name or Material 7-Bromoheptanenitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.