Learn More
7-Bromo-4-fluoroindole, 97%, Thermo Scientific™
78.79 € - 940.00 €
Chemical Identifiers
CAS | 292636-09-0 |
---|---|
Molecular Formula | C8H5BrFN |
Molecular Weight (g/mol) | 214.037 |
MDL Number | MFCD04037874 |
InChI Key | PRXUTIXCOBFMMM-UHFFFAOYSA-N |
PubChem CID | 4342956 |
IUPAC Name | 7-bromo-4-fluoro-1H-indole |
SMILES | C1=CC(=C2C(=C1F)C=CN2)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
15430698
|
Thermo Scientific Alfa Aesar
H54387.MD |
250 mg | N/A | N/A | N/A | |||||
15410698
|
Thermo Scientific Alfa Aesar
H54387.03 |
1 g | N/A | N/A | N/A | |||||
15420698
|
Thermo Scientific Alfa Aesar
H54387.06 |
5 g | N/A | N/A | N/A | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
292636-09-0 | |
214.037 | |
PRXUTIXCOBFMMM-UHFFFAOYSA-N | |
7-bromo-4-fluoro-1H-indole |
C8H5BrFN | |
MFCD04037874 | |
4342956 | |
C1=CC(=C2C(=C1F)C=CN2)Br |
Specifications
292636-09-0 | |
MFCD04037874 | |
7-bromo-4-fluoroindole, 4-fluoro-7-bromoindole, pubchem14798, 1h-indole,7-bromo-4-fluoro, 1h-indole, 7-bromo-4-fluoro, ablock ab-12-9428, 7-bromanyl-4-fluoranyl-1h-indole | |
C1=CC(=C2C(=C1F)C=CN2)Br | |
214.037 | |
214.04 | |
7-Bromo-4-fluoroindole |
C8H5BrFN | |
Air sensitive | |
PRXUTIXCOBFMMM-UHFFFAOYSA-N | |
7-bromo-4-fluoro-1H-indole | |
4342956 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Keep cold; Store under Argon
Your input is important to us. Please complete this form to provide feedback related to the content on this product.