missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Oxo-1,6-dihydropyridazine-3-carboxylic acid monohydrate, 97%, Thermo Scientific™
25.80 € - 192.00 €
Chemical Identifiers
CAS | 37972-69-3 |
---|---|
Molecular Formula | C5H3N2O3 |
Molecular Weight (g/mol) | 139.09 |
MDL Number | MFCD09064936 |
InChI Key | GIFSROMQVPUQFK-UHFFFAOYSA-M |
Synonym | 6-hydroxypyridazine-3-carboxylic acid, 6-hydroxy-3-pyridazinecarboxylic acid, 6-oxo-1,6-dihydropyridazine-3-carboxylic acid, 6-oxo-1,6-dihydro-pyridazine-3-carboxylic acid, 3-hydroxypyridazine-6-carboxylic acid, 3-pyridazinecarboxylic acid, 1,6-dihydro-6-oxo, 6-hydroxy-pyridazine-3-carboxylic acid, 6-oxo-1,6-dihydro-3-pyridazinecarboxylic acid, 3-pyridazinecarboxylicacid, 1,6-dihydro-6-oxo, 6-oxohydropyridazine-3-carboxylic acid |
PubChem CID | 305970 |
IUPAC Name | 6-oxo-1H-pyridazine-3-carboxylic acid |
SMILES | [O-]C(=O)C1=NNC(=O)C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10661473
|
Thermo Scientific Maybridge
BTB07063DA |
1 g | Amber Glass Bottle |
25.80 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10304372
|
Thermo Scientific Maybridge
BTB07063EA |
10 g | Amber Glass Bottle |
116.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10326394
|
Thermo Scientific Maybridge
BTB07063EB |
25 g | Amber Glass Bottle |
192.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
37972-69-3 | |
139.09 | |
GIFSROMQVPUQFK-UHFFFAOYSA-M | |
305970 | |
[O-]C(=O)C1=NNC(=O)C=C1 |
C5H3N2O3 | |
MFCD09064936 | |
6-hydroxypyridazine-3-carboxylic acid, 6-hydroxy-3-pyridazinecarboxylic acid, 6-oxo-1,6-dihydropyridazine-3-carboxylic acid, 6-oxo-1,6-dihydro-pyridazine-3-carboxylic acid, 3-hydroxypyridazine-6-carboxylic acid, 3-pyridazinecarboxylic acid, 1,6-dihydro-6-oxo, 6-hydroxy-pyridazine-3-carboxylic acid, 6-oxo-1,6-dihydro-3-pyridazinecarboxylic acid, 3-pyridazinecarboxylicacid, 1,6-dihydro-6-oxo, 6-oxohydropyridazine-3-carboxylic acid | |
6-oxo-1H-pyridazine-3-carboxylic acid |
Specifications
37972-69-3 | |
C5H3N2O3 | |
6-hydroxypyridazine-3-carboxylic acid, 6-hydroxy-3-pyridazinecarboxylic acid, 6-oxo-1,6-dihydropyridazine-3-carboxylic acid, 6-oxo-1,6-dihydro-pyridazine-3-carboxylic acid, 3-hydroxypyridazine-6-carboxylic acid, 3-pyridazinecarboxylic acid, 1,6-dihydro-6-oxo, 6-hydroxy-pyridazine-3-carboxylic acid, 6-oxo-1,6-dihydro-3-pyridazinecarboxylic acid, 3-pyridazinecarboxylicacid, 1,6-dihydro-6-oxo, 6-oxohydropyridazine-3-carboxylic acid | |
[O-]C(=O)C1=NNC(=O)C=C1 | |
139.09 | |
158.12 | |
6-Hydroxy-3-pyridazinecarboxylic acid monohydrate |
97% | |
MFCD09064936 | |
GIFSROMQVPUQFK-UHFFFAOYSA-M | |
6-oxo-1H-pyridazine-3-carboxylic acid | |
305970 | |
97% |
Safety and Handling
Warning