missing translation for 'onlineSavingsMsg'
Learn More

6-Morpholino-3-pyridinyl isothiocyanate, 97%, Thermo Scientific™

48.30 € - 417.00 €

Chemical Identifiers

CAS 52024-29-0
Molecular Formula C10H11N3OS
Molecular Weight (g/mol) 221.278
MDL Number MFCD03086114
InChI Key VLZWMPRZGPULIU-UHFFFAOYSA-N
Synonym 4-5-isothiocyanatopyridin-2-yl morpholine, 6-morpholino-3-pyridinyl isothiocyanate, 6-morpholino-3-pyridinylisothiocyanate, 4-5-isothiocyanato-2-pyridyl morpholine, 4-5-isothiocyatopyridin-2-yl morpholine, 6-morpholin-4-ylpyridin-3-isothiocyanate, 4-5-isothiocyanato-2-pyridinyl morpholine, 5-isothiocyanato-2-morpholin-4-yl pyridine, morpholine,4-5-isothiocyanato-2-pyridinyl, 6-morpholin-4-yl pyridin-3-yl isothiocyanate
PubChem CID 2776464
IUPAC Name 4-(5-isothiocyanatopyridin-2-yl)morpholine
SMILES C1COCCN1C2=NC=C(C=C2)N=C=S
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10089211
View Documents Promotion Details
Thermo Scientific Maybridge
CC17507CB
250 mg
48.30 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10089221
View Documents Promotion Details
Thermo Scientific Maybridge
CC17507DA
1 g
130.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
10284524
View Documents Promotion Details
Thermo Scientific Maybridge
CC17507DE
5 g
417.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

52024-29-0
221.278
VLZWMPRZGPULIU-UHFFFAOYSA-N
2776464
C1COCCN1C2=NC=C(C=C2)N=C=S
C10H11N3OS
MFCD03086114
4-5-isothiocyanatopyridin-2-yl morpholine, 6-morpholino-3-pyridinyl isothiocyanate, 6-morpholino-3-pyridinylisothiocyanate, 4-5-isothiocyanato-2-pyridyl morpholine, 4-5-isothiocyatopyridin-2-yl morpholine, 6-morpholin-4-ylpyridin-3-isothiocyanate, 4-5-isothiocyanato-2-pyridinyl morpholine, 5-isothiocyanato-2-morpholin-4-yl pyridine, morpholine,4-5-isothiocyanato-2-pyridinyl, 6-morpholin-4-yl pyridin-3-yl isothiocyanate
4-(5-isothiocyanatopyridin-2-yl)morpholine

Specifications

52024-29-0
100.0
Amber glass bottle
MFCD03086114
VLZWMPRZGPULIU-UHFFFAOYSA-N
4-(5-isothiocyanatopyridin-2-yl)morpholine
2776464
97%
96.5
97%
C10H11N3OS
4-5-isothiocyanatopyridin-2-yl morpholine, 6-morpholino-3-pyridinyl isothiocyanate, 6-morpholino-3-pyridinylisothiocyanate, 4-5-isothiocyanato-2-pyridyl morpholine, 4-5-isothiocyatopyridin-2-yl morpholine, 6-morpholin-4-ylpyridin-3-isothiocyanate, 4-5-isothiocyanato-2-pyridinyl morpholine, 5-isothiocyanato-2-morpholin-4-yl pyridine, morpholine,4-5-isothiocyanato-2-pyridinyl, 6-morpholin-4-yl pyridin-3-yl isothiocyanate
C1COCCN1C2=NC=C(C=C2)N=C=S
221.278
221.28
6-Morpholino-3-pyridinyl isothiocyanate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications