missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-1,2,3,4-tetrahydroquinoline, 98%, Thermo Scientific Chemicals

Product Code. 11442298
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

6-Methyl-1,2,3,4-tetrahydroquinoline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methyl-1,2,3,4-tetrahydroquinoline is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-61-2
Molecular Formula C10H13N
Molecular Weight (g/mol) 147.22
MDL Number MFCD00023887
InChI Key XOKMRXSMOHCNIX-UHFFFAOYSA-N
Synonym civettal, 1,2,3,4-tetrahydro-6-methylquinoline, quinoline, 1,2,3,4-tetrahydro-6-methyl, quinoline, tetrahydro-6-methyl, p-methyltetrahydroquinoline, acmc-209rck, dsstox_cid_27444, dsstox_rid_82351, dsstox_gsid_47444, xokmrxsmohcnix-uhfffaoysa
PubChem CID 66678
IUPAC Name 6-methyl-1,2,3,4-tetrahydroquinoline
SMILES CC1=CC=C2NCCCC2=C1

Specifications

Melting Point 33°C to 37°C
Boiling Point 262°C to 263°C (712 mmHg)
Quantity 10 g
Beilstein 122515
Solubility Information Insoluble in water.
Formula Weight 147.22
Percent Purity 98%
Chemical Name or Material 6-Methyl-1,2,3,4-tetrahydroquinoline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.