missing translation for 'onlineSavingsMsg'
Learn More

6-Chloroquinoline, 98%, Thermo Scientific™

Product Code. 15505076
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 612-57-7
Molecular Formula C9H6ClN
Molecular Weight (g/mol) 163.60
MDL Number MFCD00024024
InChI Key GKJSZXGYFJBYRQ-UHFFFAOYSA-N
Synonym quinoline, 6-chloro, unii-k4a4bau5se, ccris 3981, k4a4bau5se, 6-chloro-quinoline, 6-chlor-chinolin;, zlchem 764, pubchem5813, acmc-1aulp, quinoline, 6-chloro-,
PubChem CID 69163
IUPAC Name 6-chloroquinoline
SMILES ClC1=CC=C2N=CC=CC2=C1

Specifications

Melting Point 39.0°C to 42.0°C
Density 1.2700g/mL
Boiling Point 263.0°C
Flash Point 145°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Specific Gravity 1.27
Formula Weight 163.61
Percent Purity 98%
Chemical Name or Material 6-Chloroquinoline
Show More Show Less
compliance-icons
Product Identifier
  • 6-Chloroquinoline, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'