missing translation for 'onlineSavingsMsg'
Learn More

6-Chlorochroman-4-one, 99%, Thermo Scientific Chemicals

Product Code. 11447187
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

6-Chlorochroman-4-one is used as chemical, organic intermediates for pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chlorochroman-4-one is used as chemical, organic intermediates for pharmaceuticals.

Solubility
Soluble in solvents.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37674-72-9
Molecular Formula C9H7ClO2
Molecular Weight (g/mol) 182.603
MDL Number MFCD00024070
InChI Key LLTDYHFVIVSQPJ-UHFFFAOYSA-N
Synonym 6-chlorochroman-4-one, 6-chloro-4-chromanone, 4h-1-benzopyran-4-one, 6-chloro-2,3-dihydro, 6-chloro chroman-4-one, 6-chloro-2,3-dihydro-4h-chromen-4-one, 6-chloro-2,3-dihydro-1-benzopyran-4-one, 6-chloro-3,4-dihydro-2h-1-benzopyran-4-one, pubchem17931, 6-chloro-chroman-4-one, acmc-1acq5
PubChem CID 95273
IUPAC Name 6-chloro-2,3-dihydrochromen-4-one
SMILES C1COC2=C(C1=O)C=C(C=C2)Cl

Specifications

Melting Point 101°C to 102°C
Quantity 1 g
Beilstein 124218
Solubility Information Soluble in solvents.
Formula Weight 182.61
Percent Purity 99%
Chemical Name or Material 6-Chlorochroman-4-one

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.