missing translation for 'onlineSavingsMsg'
Learn More

6-(Bromomethyl)-4-chloro-2-(trifluoromethyl)quinoline, ≥95%, Thermo Scientific™

45.10 € - 330.00 €

Chemical Identifiers

CAS 123637-51-4
Molecular Formula C11H6BrClF3N
Molecular Weight (g/mol) 324.525
MDL Number MFCD00153079
InChI Key SJBKLFYWXYFAGT-UHFFFAOYSA-N
PubChem CID 2736346
IUPAC Name 6-(bromomethyl)-4-chloro-2-(trifluoromethyl)quinoline
SMILES C1=CC2=C(C=C1CBr)C(=CC(=N2)C(F)(F)F)Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10121222
View Documents
Thermo Scientific Maybridge
KM07330DA
1 g Amber Glass Bottle
45.10 €
1g
Please to purchase this item. Need a web account? Register with us today!
10314632
View Documents
Thermo Scientific Maybridge
KM07330EA
10 g Amber Glass Bottle
330.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

123637-51-4
324.525
SJBKLFYWXYFAGT-UHFFFAOYSA-N
6-(bromomethyl)-4-chloro-2-(trifluoromethyl)quinoline
C11H6BrClF3N
MFCD00153079
2736346
C1=CC2=C(C=C1CBr)C(=CC(=N2)C(F)(F)F)Cl

Specifications

123637-51-4
95%
MFCD00153079
SJBKLFYWXYFAGT-UHFFFAOYSA-N
6-(bromomethyl)-4-chloro-2-(trifluoromethyl)quinoline
2736346
95%
95.0
C11H6BrClF3N
6-bromomethyl-4-chloro-2-trifluoromethyl quinoline, quinoline,6-bromomethyl-4-chloro-2-trifluoromethyl, quinoline, 6-bromomethyl-4-chloro-2-trifluoromethyl, pubchem6004, acmc-1bzc1, buttpark 29\04-10, 6-bromomethyl-4-chloro-2-trifluoromethyl-quinoline, 2-trifluoromethyl-4-chloro-6-bromomethyl quinoline
C1=CC2=C(C=C1CBr)C(=CC(=N2)C(F)(F)F)Cl
324.525
324.53
6-(Bromomethyl)-4-chloro-2-(trifluoromethyl)quinoline
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.