missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-benzothiazolinone, 98%, Thermo Scientific™

Product Code. 15466848
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62266-82-4
Molecular Formula C7H4BrNOS
Molecular Weight (g/mol) 230.08
MDL Number MFCD00239363
InChI Key HECJMTPEVWQFCY-UHFFFAOYSA-N
Synonym 6-bromo-2-benzothiazolinone, 6-bromobenzo d thiazol-2 3h-one, 6-bromo-2 3h-benzothiazolone, 6-bromo-3h-benzothiazol-2-one, 2 3h-benzothiazolone, 6-bromo, 6-bromo-1,3-benzothiazol-2 3h-one, 6-bromo-2-hydroxy benzothiazol, 6-bromo-2,3-dihydro-1,3-benzothiazol-2-one, 2-benzothiazolol, 6-bromo, pubchem21839
PubChem CID 188444
SMILES BrC1=CC=C2NC(=O)SC2=C1

Specifications

Melting Point 229°C to 233°C
Quantity 5 g
IUPAC Name 6-bromo-2,3-dihydro-1,3-benzothiazol-2-one
Formula Weight 230.08
Percent Purity 98%
Chemical Name or Material 6-Bromo-2-benzothiazolinone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'