missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2,4-dichloroquinazoline, 97%, Thermo Scientific Chemicals

56.90 € - 824.00 €

Chemical Identifiers

CAS 102393-82-8
Molecular Formula C8H3BrCl2N2
Molecular Weight (g/mol) 277.93
MDL Number MFCD09744007
InChI Key LBAYOWRVZAKPLS-UHFFFAOYSA-N
PubChem CID 10107568
IUPAC Name 6-bromo-2,4-dichloroquinazoline
SMILES C1=CC2=C(C=C1Br)C(=NC(=N2)Cl)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15410389
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H63603.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15490379
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H63603.03
1 g
182.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
15400389
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H63603.06
5 g
824.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

102393-82-8
277.93
LBAYOWRVZAKPLS-UHFFFAOYSA-N
6-bromo-2,4-dichloroquinazoline
C8H3BrCl2N2
MFCD09744007
10107568
C1=CC2=C(C=C1Br)C(=NC(=N2)Cl)Cl

Specifications

102393-82-8
C8H3BrCl2N2
quinazoline, 6-bromo-2,4-dichloro, 2,4-dichloro-6-bromoquinazoline, 6-bromo-2,4-dichloro-quinazoline, quinazoline,6-bromo-2,4-dichloro, pubchem14690, acmc-1c6nj, abbypharma ap-16-10560, 6-bromanyl-2,4-bis chloranyl quinazoline
LBAYOWRVZAKPLS-UHFFFAOYSA-N
6-bromo-2,4-dichloroquinazoline
10107568
97%
6-Bromo-2,4-dichloroquinazoline
White
MFCD09744007
Slightly soluble in water.
C1=CC2=C(C=C1Br)C(=NC(=N2)Cl)Cl
277.93
277.93
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only