missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-7-(5-indolinyl)-9-methyl-7-deazapurine, 95%, Thermo Scientific Chemicals

Product Code. 15495096
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1337596-50-5
Molecular Formula C15H15N5
Molecular Weight (g/mol) 265.32
MDL Number MFCD22988986
InChI Key CXRNQXFFWARLSJ-UHFFFAOYSA-N
Synonym 6-amino-7-5-indolinyl-9-methyl-7-deazapurine, 5-2,3-dihydro-1h-indol-5-yl-7-methylpyrrolo 2,3-d pyrimidin-4-amine, 5-2,3-dihydro-1h-indol-5-yl-7-methyl-7h-pyrrolo 2,3-d pyrimidin-4-amine
PubChem CID 66823471
IUPAC Name 5-(2,3-dihydro-1H-indol-5-yl)-7-methylpyrrolo[2,3-d]pyrimidin-4-amine
SMILES CN1C=C(C2=C1N=CN=C2N)C3=CC4=C(C=C3)NCC4

Specifications

Quantity 250 mg
Formula Weight 265.31
Percent Purity 95%
Chemical Name or Material 6-Amino-7-(5-indolinyl)-9-methyl-7-deazapurine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'