Learn More
6,7-Dihydrobenzo[b]thiophen-4-one, 98%, Thermo Scientific Chemicals
114.57 € - 441.20 €
Chemical Identifiers
CAS | 13414-95-4 |
---|---|
Molecular Formula | C8H8OS |
Molecular Weight (g/mol) | 152.211 |
MDL Number | MFCD00005861 |
InChI Key | GJEKNELSXNSYAQ-UHFFFAOYSA-N |
Synonym | 6,7-dihydro-4-benzo b thiophenone, 6,7-dihydro-5h-benzo b thiophen-4-one, 6,7-dihydrobenzo b thiophen-4 5h-one, 4-keto-4,5,6,7-tetrahydrothianaphthene, 6,7-dihydrobenzo b thiophen-4-one, benzo b thiophen-4 5h-one, 6,7-dihydro, 6,7-dihydro-1-benzothiophen-4 5h-one, 4,5,6,7-tetrahydrothionaphthen-4-one, 4-oxo-4,5,6,7-tetrahydrothianaphthene, 4,5,6,7-tetrahydro-4-benzothiophenone |
PubChem CID | 83418 |
IUPAC Name | 6,7-dihydro-5H-1-benzothiophen-4-one |
SMILES | C1CC2=C(C=CS2)C(=O)C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15446698
|
Thermo Scientific Alfa Aesar
H60220.04 |
2 g |
114.57 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15456698
|
Thermo Scientific Alfa Aesar
H60220.09 |
10 g |
441.20 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.
Solubility
Soluble in chloroform.
Notes
Store at room temperature. Keep away from oxidizing agents.
Chemical Identifiers
13414-95-4 | |
152.211 | |
GJEKNELSXNSYAQ-UHFFFAOYSA-N | |
83418 | |
C1CC2=C(C=CS2)C(=O)C1 |
C8H8OS | |
MFCD00005861 | |
6,7-dihydro-4-benzo b thiophenone, 6,7-dihydro-5h-benzo b thiophen-4-one, 6,7-dihydrobenzo b thiophen-4 5h-one, 4-keto-4,5,6,7-tetrahydrothianaphthene, 6,7-dihydrobenzo b thiophen-4-one, benzo b thiophen-4 5h-one, 6,7-dihydro, 6,7-dihydro-1-benzothiophen-4 5h-one, 4,5,6,7-tetrahydrothionaphthen-4-one, 4-oxo-4,5,6,7-tetrahydrothianaphthene, 4,5,6,7-tetrahydro-4-benzothiophenone | |
6,7-dihydro-5H-1-benzothiophen-4-one |
Specifications
13414-95-4 | |
120°C | |
C8H8OS | |
MFCD00005861 | |
6,7-dihydro-4-benzo b thiophenone, 6,7-dihydro-5h-benzo b thiophen-4-one, 6,7-dihydrobenzo b thiophen-4 5h-one, 4-keto-4,5,6,7-tetrahydrothianaphthene, 6,7-dihydrobenzo b thiophen-4-one, benzo b thiophen-4 5h-one, 6,7-dihydro, 6,7-dihydro-1-benzothiophen-4 5h-one, 4,5,6,7-tetrahydrothionaphthen-4-one, 4-oxo-4,5,6,7-tetrahydrothianaphthene, 4,5,6,7-tetrahydro-4-benzothiophenone | |
GJEKNELSXNSYAQ-UHFFFAOYSA-N | |
6,7-dihydro-5H-1-benzothiophen-4-one | |
83418 | |
98% |
38°C to 40°C | |
110°C (230°F) | |
1.5895 | |
112276 | |
Soluble in chloroform. | |
C1CC2=C(C=CS2)C(=O)C1 | |
152.211 | |
152.21 | |
6,7-Dihydrobenzo[b]thiophen-4-one |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 236-510-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.