missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-(2-furyl)nicotinic acid, Thermo Scientific™
83.70 € - 695.00 €
Chemical Identifiers
CAS | 884507-36-2 |
---|---|
Molecular Formula | C10H7NO3 |
Molecular Weight (g/mol) | 189.17 |
MDL Number | MFCD06410385 |
InChI Key | ALJRNVXLRKHVQF-UHFFFAOYSA-N |
Synonym | 6-2-furyl nicotinic acid, 6-furan-2-yl nicotinic acid, 6-furan-2-yl pyridine-3-carboxylic acid, 6-2-furyl pyridine-3-carboxylic acid |
PubChem CID | 19010290 |
IUPAC Name | 6-(furan-2-yl)pyridine-3-carboxylic acid |
SMILES | OC(=O)C1=CN=C(C=C1)C1=CC=CO1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10001934
|
Thermo Scientific Maybridge
CC54901CB |
250 mg |
83.70 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10144913
|
Thermo Scientific Maybridge
CC54901DA |
1 g |
227.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10557484
|
Thermo Scientific Maybridge
CC54901DE |
5 g |
695.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
884507-36-2 | |
189.17 | |
ALJRNVXLRKHVQF-UHFFFAOYSA-N | |
19010290 | |
OC(=O)C1=CN=C(C=C1)C1=CC=CO1 |
C10H7NO3 | |
MFCD06410385 | |
6-2-furyl nicotinic acid, 6-furan-2-yl nicotinic acid, 6-furan-2-yl pyridine-3-carboxylic acid, 6-2-furyl pyridine-3-carboxylic acid | |
6-(furan-2-yl)pyridine-3-carboxylic acid |
Specifications
884507-36-2 | |
C10H7NO3 | |
6-2-furyl nicotinic acid, 6-furan-2-yl nicotinic acid, 6-furan-2-yl pyridine-3-carboxylic acid, 6-2-furyl pyridine-3-carboxylic acid | |
OC(=O)C1=CN=C(C=C1)C1=CC=CO1 | |
189.17 | |
189.17 | |
6-(2-Furyl)nicotinic acid |
97% | |
MFCD06410385 | |
ALJRNVXLRKHVQF-UHFFFAOYSA-N | |
6-(furan-2-yl)pyridine-3-carboxylic acid | |
19010290 | |
97% |
Safety and Handling
GHS Signal Word: Warning
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title