Learn More
5Z-7-Oxozeaenol, Thermo Scientific Chemicals
97.70 € - 474.00 €
Chemical Identifiers
CAS | 66018-38-0 |
---|---|
Molecular Formula | C19H22O7 |
Molecular Weight (g/mol) | 362.378 |
MDL Number | MFCD12912700 |
InChI Key | NEQZWEXWOFPKOT-UHFFFAOYSA-N |
Synonym | 8,9,16-trihydroxy-14-methoxy-3-methyl-4,8,9,10-tetrahydro-3h-2-benzoxacyclotetradecine-1,7-dione |
PubChem CID | 53442201 |
IUPAC Name | 5,6,15-trihydroxy-17-methoxy-11-methyl-12-oxabicyclo[12.4.0]octadeca-1(14),2,8,15,17-pentaene-7,13-dione |
SMILES | CC1CC=CC(=O)C(C(CC=CC2=CC(=CC(=C2C(=O)O1)O)OC)O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15428099
|
Thermo Scientific Alfa Aesar
J66651.M^ |
1 mg |
97.70 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15418099
|
Thermo Scientific Alfa Aesar
J66651.#0 |
5 mg |
474.00 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
66018-38-0 | |
362.378 | |
NEQZWEXWOFPKOT-UHFFFAOYSA-N | |
53442201 | |
CC1CC=CC(=O)C(C(CC=CC2=CC(=CC(=C2C(=O)O1)O)OC)O)O |
C19H22O7 | |
MFCD12912700 | |
8,9,16-trihydroxy-14-methoxy-3-methyl-4,8,9,10-tetrahydro-3h-2-benzoxacyclotetradecine-1,7-dione | |
5,6,15-trihydroxy-17-methoxy-11-methyl-12-oxabicyclo[12.4.0]octadeca-1(14),2,8,15,17-pentaene-7,13-dione |
Specifications
66018-38-0 | |
C19H22O7 | |
UN2811 | |
Soluble in DMSO | |
CC1CC=CC(=O)C(C(CC=CC2=CC(=CC(=C2C(=O)O1)O)OC)O)O | |
362.378 | |
362.37 | |
5Z-7-Oxozeaenol |
White | |
MFCD12912700 | |
8,9,16-trihydroxy-14-methoxy-3-methyl-4,8,9,10-tetrahydro-3h-2-benzoxacyclotetradecine-1,7-dione | |
NEQZWEXWOFPKOT-UHFFFAOYSA-N | |
5,6,15-trihydroxy-17-methoxy-11-methyl-12-oxabicyclo[12.4.0]octadeca-1(14),2,8,15,17-pentaene-7,13-dione | |
53442201 | |
Solid |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'tsca' : No
missing translation for 'storageNote' : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only