Learn More
Spectrum™ Labs Spectra/Por™ 3 3.5 kD MWCO Standard RC Dry Dialysis Kits
With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated
Marca: Spectrum™ 132720T
150.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Includes:
- 1m or 5m of Spectra/Por 3 dialysis tubing
- 1 Standard Spectra/Por Closure (for buoyance)
- 1 weighted Spectra/Por Closure (for orientation)
- 2 adhesive tubing openers
Descripción
- For basic dialysis applications, or evaluating dialysis tubing for dialysis application
- Packaged dry with glycerin (soak in water) or without glycerin (no soaking required)
Especificaciones
3.5 kDa | |
11.5 mm | |
5m roll of Spectra/Por™ 1, 2 or 3 dialysis tubing, 1 standard spectra/por closure (aids buoyancy), 1 weighted spectra/por closure (aids vertical orientation), 5 opening picks | |
Ideal for small volume dialysis or evaluation of membrane for an application |
Regenerated Cellulose | |
1.1 mL/cm vol./length | |
5 m Tubing/Kit |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido