Learn More
Hamilton™ 500 μL Microliter Syringe Model 750 N, Cemented Needle, 22 gauge, 2 in, point style 2
Split/splitless injection consists of a heated chamber with a glass liner into which the sample is injectedthrough the septum. A microsyringe is used to inject the sample through a rubber septum into a flash vaporizer chamber at the head of the column.
Brand: Hamilton™ DS80800
Description
Specification Sheet
- Volume – 500 μL
- Syringe Type – Microliter
- Syringe Series – 700
- Termination – Cemented Needle (N)
- Needle – Cemented Needle
- Gauge – 22 gauge
- Point Style – 2
- Needle Length – 2 inch (51 mm)
- Fluid Path – SST, borosilicate glass
- Autoclavable – No
- Gas Sterilizable – Yes
- Pressure – 1000 psig (68.9 bar)
- Maximum Temperature – 50°C / 122°F
- Minimum Temperature – 10°C / 50°F
- Barrel Inner Diameter – 0.135 inches (3.26 mm)
- Barrel Outer Diameter – 0.305 inches (7.75 mm)
- Needle Inner Diameter – 0.0161 inches (0.41 mm)
- Needle Outer Diameter – 0.0283 inches (0.72 mm)
- Std Needle Dead Volume – 6.81 μL
- Sales Quantity – 1 syringe per pack
Specifications
22 G | |
700 series | |
500 μL | |
51 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.