missing translation for 'onlineSavingsMsg'
Learn More

5-Pyrimidinylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 15465315
Click to view available options
Quantity:
50 mL
Packaging:
ChemSeal™ bottles
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Reacts with water.

Notes
Air Sensitive. Store under dry inert gas. Store away from air. Incompatible with oxidizing agents, acid chlorides. Protect from humidity and water. Do not together with acids.
TRUSTED_SUSTAINABILITY

Especificaciones

CAS 1227941-02-7
Color Black to Yellow
Packaging ChemSeal™ bottles
Molecular Formula C4H3BrN2Zn
MDL Number MFCD11915900
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive; May form a precipitate
Synonym 5-pyrimidylzinc bromide, 5-pyrimidinylzinc bromide, 0.5m in thf, pyrimidin-5-ylzinc bromide, 0.50 m in thf, 5-pyrimidylzinc bromide 0.5 m in tetrahydrofuran, 5-pyrimidylzinc bromide, 0.5m in tetrahydrofuran, 5-pyrimidinylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
Solubility Information Reacts with water.
InChI Key QWKPLWAOLNGTMA-UHFFFAOYSA-M
SMILES C1=[C-]C=NC=N1.[Zn+]Br
IUPAC Name bromozinc(1+);5H-pyrimidin-5-ide
Molecular Weight (g/mol) 224.366
PubChem CID 49761916
Formula Weight 224.37
Physical Form Liquid
Chemical Name or Material 5-Pyrimidinylzinc bromide
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado