missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Piperidinothiophene-2-carbaldehyde, 97%, Thermo Scientific™
32.20 € - 78.90 €
Chemical Identifiers
CAS | 24372-48-3 |
---|---|
Molecular Formula | C10H13NOS |
Molecular Weight (g/mol) | 195.28 |
MDL Number | MFCD05263177 |
InChI Key | SFDXRMCPPLZJBG-UHFFFAOYSA-N |
PubChem CID | 2113341 |
SMILES | O=CC1=CC=C(S1)N1CCCCC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10778435
|
Thermo Scientific Maybridge
CC72604CB |
250 mg | Amber Glass Bottle |
32.20 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10547084
|
Thermo Scientific Maybridge
CC72604DA |
1 g | Amber Glass Bottle |
78.90 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
24372-48-3 | |
195.28 | |
SFDXRMCPPLZJBG-UHFFFAOYSA-N | |
O=CC1=CC=C(S1)N1CCCCC1 |
C10H13NOS | |
MFCD05263177 | |
2113341 |
Specifications
24372-48-3 | |
100.0 | |
C10H13NOS | |
5-piperidin-1-yl thiophene-2-carbaldehyde, 5-piperidin-1-yl-thiophene-2-carbaldehyde, 5-piperidino-2-thiophenecarbaldehyde, 5-piperidin-1-yl-thiophene-2-carboxaldehyde, 5-piperidylthiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde, 5-1-piperidinyl, piperidinothiophenecarbaldehyde, 5-piperidin-1-yl-2-formylthiophene, 2-thiophenecarboxaldehyde,5-1-piperidinyl | |
O=CC1=CC=C(S1)N1CCCCC1 | |
195.28 | |
195.28 | |
5-Piperidinothiophene-2-carbaldehyde |
96.5 | |
97% | |
MFCD05263177 | |
SFDXRMCPPLZJBG-UHFFFAOYSA-N | |
5-piperidin-1-ylthiophene-2-carbaldehyde | |
2113341 | |
97% |