missing translation for 'onlineSavingsMsg'
Learn More

5-Phenyl-2-furaldehyde, 96%, Thermo Scientific Chemicals

Product Code. 10844531
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13803-39-9
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.19
InChI Key BMJHNNPEPBZULA-UHFFFAOYSA-N
Synonym 5-phenyl-2-furaldehyde, 2-formyl-5-phenylfuran, 5-phenyl-furan-2-carbaldehyde, 5-phenylfurfural, 2-furancarboxaldehyde,5-phenyl, 5-phenylfuraldehyde, acmc-209y7p, 5-phenyl-2-furancarboxaldehyde
PubChem CID 2769420
IUPAC Name 5-phenylfuran-2-carbaldehyde
SMILES C1=CC=C(C=C1)C2=CC=C(O2)C=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 29°C to 33°C
Color Brown to Yellow
Boiling Point 104°C to 108°C (6.0 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 172.19
Percent Purity 96%
Physical Form Liquid or Solid
Chemical Name or Material 5-Phenyl-2-furaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 5-Phenyl-2-furaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.