Learn More
5-Nitroisatin, 97%, Thermo Scientific Chemicals
40.70 € - 121.00 €
Chemical Identifiers
CAS | 611-09-6 |
---|---|
Molecular Formula | C8H4N2O4 |
Molecular Weight (g/mol) | 192.13 |
MDL Number | MFCD00005720 |
InChI Key | UNMYHYODJHKLOC-UHFFFAOYSA-N |
Synonym | 5-nitroisatin, 5-nitroindoline-2,3-dione, 5-nitroindole-2,3-dione, 1h-indole-2,3-dione, 5-nitro, indole-2,3-dione, 5-nitro, isatin, 5-nitro, n-nitroisatin, ccris 4031, 2,3-dihydro-5-nitroindole-2,3-dione, 5-nitro-2,3-dihydro-1h-indole-2,3-dione |
PubChem CID | 4669250 |
IUPAC Name | 5-nitro-1H-indole-2,3-dione |
SMILES | C1=CC2=C(C=C1[N+](=O)[O-])C(=O)C(=O)N2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10785401
|
Thermo Scientific Acros
128720050 |
5 g | Glass bottle |
40.70 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10031430
|
Thermo Scientific Acros
128720250 |
25 g | Glass bottle |
121.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
611-09-6 | |
192.13 | |
UNMYHYODJHKLOC-UHFFFAOYSA-N | |
4669250 | |
C1=CC2=C(C=C1[N+](=O)[O-])C(=O)C(=O)N2 |
C8H4N2O4 | |
MFCD00005720 | |
5-nitroisatin, 5-nitroindoline-2,3-dione, 5-nitroindole-2,3-dione, 1h-indole-2,3-dione, 5-nitro, indole-2,3-dione, 5-nitro, isatin, 5-nitro, n-nitroisatin, ccris 4031, 2,3-dihydro-5-nitroindole-2,3-dione, 5-nitro-2,3-dihydro-1h-indole-2,3-dione | |
5-nitro-1H-indole-2,3-dione |
Specifications
611-09-6 | |
100.0 | |
Orange to Orange-Yellow | |
97% | |
MFCD00005720 | |
5-nitroisatin, 5-nitroindoline-2,3-dione, 5-nitroindole-2,3-dione, 1h-indole-2,3-dione, 5-nitro, indole-2,3-dione, 5-nitro, isatin, 5-nitro, n-nitroisatin, ccris 4031, 2,3-dihydro-5-nitroindole-2,3-dione, 5-nitro-2,3-dihydro-1h-indole-2,3-dione | |
C1=CC2=C(C=C1[N+](=O)[O-])C(=O)C(=O)N2 | |
192.13 | |
192.13 | |
Crystalline Powder |
96.0 | |
251°C | |
Authentic | |
C8H4N2O4 | |
21, 456 | |
UNMYHYODJHKLOC-UHFFFAOYSA-N | |
5-nitro-1H-indole-2,3-dione | |
4669250 | |
97% | |
5-Nitroisatin, 97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Suspected of causing genetic defects.
GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 210-252-5
missing translation for 'rtecsNumber' : NL7970000
- 5-Nitroisatin
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Germ cell mutagenicity Category 2
Hazard Statement
- H341-Suspected of causing genetic defects.
- H302-Harmful if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.