Learn More
5-Methyl-2-furaldehyde, 98%, Thermo Scientific Chemicals
36.68 € - 351.00 €
Chemical Identifiers
CAS | 620-02-0 |
---|---|
Molecular Formula | C6H6O2 |
Molecular Weight (g/mol) | 110.112 |
MDL Number | MFCD00003232 |
InChI Key | OUDFNZMQXZILJD-UHFFFAOYSA-N |
Synonym | 5-methylfurfural, 5-methyl-2-furaldehyde, 5-methyl furfural, 5-methyl-2-furfural, 2-furancarboxaldehyde, 5-methyl, 5-methylfuran-2-al, 5-methyl-2-furancarboxaldehyde, 5-methylfurfuraldehyde, 2-furaldehyde, 5-methyl, 2-formyl-5-methylfuran |
PubChem CID | 12097 |
ChEBI | CHEBI:2091 |
IUPAC Name | 5-methylfuran-2-carbaldehyde |
SMILES | CC1=CC=C(O1)C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11490803
|
Thermo Scientific Alfa Aesar
A13264.14 |
25 g |
36.68 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11400813
|
Thermo Scientific Alfa Aesar
A13264.22 |
100 g |
100.50 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11410813
|
Thermo Scientific Alfa Aesar
A13264.36 |
500 g | N/A | ||||||
Description
5-Methyl-2-furaldehyde is used as a perfuming agent. It is used a smoke with essence.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Methyl-2-furaldehyde is used as a perfuming agent. It is used a smoke with essence.
Solubility
Soluble in alcohol and water.
Notes
Store at room temperature. Incompatible with oxidizing agents. Air sensitive.
Chemical Identifiers
620-02-0 | |
110.112 | |
OUDFNZMQXZILJD-UHFFFAOYSA-N | |
12097 | |
5-methylfuran-2-carbaldehyde |
C6H6O2 | |
MFCD00003232 | |
5-methylfurfural, 5-methyl-2-furaldehyde, 5-methyl furfural, 5-methyl-2-furfural, 2-furancarboxaldehyde, 5-methyl, 5-methylfuran-2-al, 5-methyl-2-furancarboxaldehyde, 5-methylfurfuraldehyde, 2-furaldehyde, 5-methyl, 2-formyl-5-methylfuran | |
CHEBI:2091 | |
CC1=CC=C(O1)C=O |
Specifications
620-02-0 | |
186°C to 188°C | |
C6H6O2 | |
MFCD00003232 | |
Air Sensitive | |
Soluble in alcohol and water. | |
CC1=CC=C(O1)C=O | |
110.112 | |
CHEBI:2091 | |
98% |
1.106 | |
72°C (161°F) | |
1.531 | |
106895 | |
5-methylfurfural, 5-methyl-2-furaldehyde, 5-methyl furfural, 5-methyl-2-furfural, 2-furancarboxaldehyde, 5-methyl, 5-methylfuran-2-al, 5-methyl-2-furancarboxaldehyde, 5-methylfurfuraldehyde, 2-furaldehyde, 5-methyl, 2-formyl-5-methylfuran | |
OUDFNZMQXZILJD-UHFFFAOYSA-N | |
5-methylfuran-2-carbaldehyde | |
12097 | |
110.11 | |
5-Methyl-2-furaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319
missing translation for 'einecsNumber' : 210-622-6
missing translation for 'rtecsNumber' : LT7032500
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.