missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyisatin, 97%, Thermo Scientific Chemicals

Product Code. 13555030
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39755-95-8
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.16
MDL Number MFCD00169023
InChI Key DMHGXMPXHPOXBF-UHFFFAOYSA-N
Synonym 5-methoxyisatin, 5-methoxyindoline-2,3-dione, 5-methoxy-2,3-indolinedione, 1h-indole-2,3-dione, 5-methoxy, 5-methoxy-indole-2,3-dione, 5-methoxy isatin, 5-methoxyindole-2,3-dione, 5-methoxy-2,3-dihydro-1h-indole-2,3-dione, indole-2,3-dione, 5-methoxy, 1h-indole-2,3-dione, 5-methoxy-9ci
PubChem CID 38333
IUPAC Name 5-methoxy-1H-indole-2,3-dione
SMILES COC1=CC=C2NC(=O)C(=O)C2=C1

Specifications

Melting Point 201°C to 203°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 177.16
Percent Purity 97%
Chemical Name or Material 5-Methoxyisatin
compliance-icons
Product Identifier
  • 5-Methoxyisatin
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'