Learn More
5-Iodoindole, 95%, Thermo Scientific Chemicals
84.10 € - 263.00 €
Chemical Identifiers
CAS | 16066-91-4 |
---|---|
Molecular Formula | C8H6IN |
Molecular Weight (g/mol) | 243.04 |
InChI Key | TVQLYTUWUQMGMP-UHFFFAOYSA-N |
Synonym | 5-iodoindole, 1h-indole, 5-iodo, 5-iodo indole, 5-iodanyl-1h-indole, zlchem 1099, acmc-1brxb, maybridge1_006499 |
PubChem CID | 2782313 |
IUPAC Name | 5-iodo-1H-indole |
SMILES | C1=CC2=C(C=CN2)C=C1I |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11986721
|
Thermo Scientific Acros
441590050 |
5 g |
84.10 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11996721
|
Thermo Scientific Acros
441590250 |
25 g |
263.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16066-91-4 | |
243.04 | |
5-iodoindole, 1h-indole, 5-iodo, 5-iodo indole, 5-iodanyl-1h-indole, zlchem 1099, acmc-1brxb, maybridge1_006499 | |
5-iodo-1H-indole |
C8H6IN | |
TVQLYTUWUQMGMP-UHFFFAOYSA-N | |
2782313 | |
C1=CC2=C(C=CN2)C=C1I |
Specifications
16066-91-4 | |
100.0 | |
Brown to White | |
94% min. (HPLC) | |
5-iodoindole, 1h-indole, 5-iodo, 5-iodo indole, 5-iodanyl-1h-indole, zlchem 1099, acmc-1brxb, maybridge1_006499 | |
C1=CC2=C(C=CN2)C=C1I | |
243.04 | |
243.04 | |
Crystalline Solid or Solid |
94.0 | |
95°C to 99°C | |
Authentic | |
C8H6IN | |
TVQLYTUWUQMGMP-UHFFFAOYSA-N | |
5-iodo-1H-indole | |
2782313 | |
95% | |
5-Iodoindole |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
Warning
- 5-Iodoindole
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only