missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyindole-3-acetic Acid, 99%, Thermo Scientific Chemicals

75.98 € - 649.23 €

Chemical Identifiers

CAS 54-16-0
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.19
MDL Number MFCD00005639
InChI Key DUUGKQCEGZLZNO-UHFFFAOYSA-N
Synonym 5-hydroxyindole-3-acetic acid, 5-hiaa, 2-5-hydroxy-1h-indol-3-yl acetic acid, 5-hydroxyindoleacetic acid, 5-hydroxyheteroauxin, 5-hydroxy-1h-indol-3-yl acetic acid, 5-oxyindoleacetic acid, hydroxyindoleacetic acid, 5-hydroxy-iaa, 5-hydroxyindol-3-ylacetic acid
PubChem CID 1826
ChEBI CHEBI:27823
IUPAC Name 2-(5-hydroxy-1H-indol-3-yl)acetic acid
SMILES C1=CC2=C(C=C1O)C(=CN2)CC(=O)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10235160
SDS View Documents
Thermo Scientific Acros
121282500
250 mg Glass bottle
75.98 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10793861
SDS View Documents
Thermo Scientific Acros
121280010
1 g Glass bottle
203.01 €
1g
Please to purchase this item. Need a web account? Register with us today!
10524074
SDS View Documents
Thermo Scientific Acros
121280050
5 g Glass bottle
649.23 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

54-16-0
191.19
DUUGKQCEGZLZNO-UHFFFAOYSA-N
1826
2-(5-hydroxy-1H-indol-3-yl)acetic acid
C10H9NO3
MFCD00005639
5-hydroxyindole-3-acetic acid, 5-hiaa, 2-5-hydroxy-1h-indol-3-yl acetic acid, 5-hydroxyindoleacetic acid, 5-hydroxyheteroauxin, 5-hydroxy-1h-indol-3-yl acetic acid, 5-oxyindoleacetic acid, hydroxyindoleacetic acid, 5-hydroxy-iaa, 5-hydroxyindol-3-ylacetic acid
CHEBI:27823
C1=CC2=C(C=C1O)C(=CN2)CC(=O)O

Specifications

54-16-0
100.0
Beige
99%
MFCD00005639
Solubility in water: slightly soluble
C1=CC2=C(C=C1O)C(=CN2)CC(=O)O
191.19
CHEBI:27823
99%
5-Hydroxyindole-3-acetic acid
98.5
162°C to 165°C
Authentic
C10H9NO3
5-hydroxyindole-3-acetic acid, 5-hiaa, 2-5-hydroxy-1h-indol-3-yl acetic acid, 5-hydroxyindoleacetic acid, 5-hydroxyheteroauxin, 5-hydroxy-1h-indol-3-yl acetic acid, 5-oxyindoleacetic acid, hydroxyindoleacetic acid, 5-hydroxy-iaa, 5-hydroxyindol-3-ylacetic acid
DUUGKQCEGZLZNO-UHFFFAOYSA-N
2-(5-hydroxy-1H-indol-3-yl)acetic acid
1826
191.19
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 200-195-4

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Hydroxyindole-3-acetic Acid, 99%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.