missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Hydroxy-2-adamantone, 98%, Thermo Scientific™
18.80 € - 274.00 €
Chemical Identifiers
CAS | 20098-14-0 |
---|---|
Molecular Formula | C10H14O2 |
Molecular Weight (g/mol) | 166.22 |
MDL Number | MFCD00192211 |
InChI Key | TZBDEVBNMSLVKT-UHFFFAOYSA-N |
Synonym | idramantone, 5-hydroxy-2-adamantanone, kemantane, 5-hydroxy-adamantan-2-one, kemantan, 5-hydroxytricyclo 3.3.1.1 3,7 decanone, tricyclo 3.3.1.13,7 decanone, 5-hydroxy, idramantone inn, 4-oxo-1-adamantanol |
PubChem CID | 64184 |
ChEBI | CHEBI:48581 |
IUPAC Name | 5-hydroxyadamantan-2-one |
SMILES | C1C2CC3CC(C2)(CC1C3=O)O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
20098-14-0 | |
166.22 | |
TZBDEVBNMSLVKT-UHFFFAOYSA-N | |
64184 | |
5-hydroxyadamantan-2-one |
C10H14O2 | |
MFCD00192211 | |
idramantone, 5-hydroxy-2-adamantanone, kemantane, 5-hydroxy-adamantan-2-one, kemantan, 5-hydroxytricyclo 3.3.1.1 3,7 decanone, tricyclo 3.3.1.13,7 decanone, 5-hydroxy, idramantone inn, 4-oxo-1-adamantanol | |
CHEBI:48581 | |
C1C2CC3CC(C2)(CC1C3=O)O |
Specifications
20098-14-0 | |
C10H14O2 | |
idramantone, 5-hydroxy-2-adamantanone, kemantane, 5-hydroxy-adamantan-2-one, kemantan, 5-hydroxytricyclo 3.3.1.1 3,7 decanone, tricyclo 3.3.1.13,7 decanone, 5-hydroxy, idramantone inn, 4-oxo-1-adamantanol | |
C1C2CC3CC(C2)(CC1C3=O)O | |
166.22 | |
CHEBI:48581 | |
98% |
>300°C | |
MFCD00192211 | |
TZBDEVBNMSLVKT-UHFFFAOYSA-N | |
5-hydroxyadamantan-2-one | |
64184 | |
166.22 | |
5-Hydroxy-2-adamantone |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title