Learn More
5-Hexynenitrile, 98%, Thermo Scientific Chemicals
29.60 € - 82.10 €
Chemical Identifiers
CAS | 14918-21-9 |
---|---|
Molecular Formula | C6H7N |
Molecular Weight (g/mol) | 93.129 |
MDL Number | MFCD00001978 |
InChI Key | JZYKFLLRVPPISG-UHFFFAOYSA-N |
Synonym | 5-hexynenitrile, 5-cyano-1-pentyne, acmc-1c81n, 5-hexynenitrile 1g |
PubChem CID | 139852 |
IUPAC Name | hex-5-ynenitrile |
SMILES | C#CCCCC#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11397755
|
Thermo Scientific Alfa Aesar
L02604.03 |
1 g |
29.60 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11307765
|
Thermo Scientific Alfa Aesar
L02604.06 |
5 g |
82.10 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
5-Hexynenitrile is used in the preparation of annelated symmetric and asymmetric 3,3'-disubstituted 2,2'-bipyridines by cobalt(I)-catalyzed [2+2+2] cycloadditions with 1,3-diynes. Further, it reacts with 3-bromo-pyridine to prepare 6-(3-pyridinyl)-5-hexynenitrile.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Hexynenitrile is used in the preparation of annelated symmetric and asymmetric 3,3′-disubstituted 2,2′-bipyridines by cobalt(I)-catalyzed [2+2+2] cycloadditions with 1,3-diynes. Further, it reacts with 3-bromo-pyridine to prepare 6-(3-pyridinyl)-5-hexynenitrile.
Solubility
Miscible with water.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
Chemical Identifiers
14918-21-9 | |
93.129 | |
JZYKFLLRVPPISG-UHFFFAOYSA-N | |
139852 | |
C#CCCCC#N |
C6H7N | |
MFCD00001978 | |
5-hexynenitrile, 5-cyano-1-pentyne, acmc-1c81n, 5-hexynenitrile 1g | |
hex-5-ynenitrile |
Specifications
14918-21-9 | |
115°C to 117°C | |
C6H7N | |
MFCD00001978 | |
1735926 | |
Miscible with water. | |
C#CCCCC#N | |
93.129 | |
93.13 | |
5-Hexynenitrile |
0.889 | |
42°C (107°F) | |
1.441 | |
UN1992 | |
5-hexynenitrile, 5-cyano-1-pentyne, acmc-1c81n, 5-hexynenitrile 1g | |
JZYKFLLRVPPISG-UHFFFAOYSA-N | |
hex-5-ynenitrile | |
139852 | |
98% |
Safety and Handling
GHS H Statement
H226-H302-H312-H332
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only