missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoroindoline, 97%, Thermo Scientific Chemicals

Product Code. 15430528
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

5-Fluoroindoline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Fluoroindoline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2343-22-8
Molecular Formula C8H8FN
Molecular Weight (g/mol) 137.157
MDL Number MFCD00214461
InChI Key NXQRMQIYCWFDGP-UHFFFAOYSA-N
PubChem CID 2774463
IUPAC Name 5-fluoro-2,3-dihydro-1H-indole
SMILES C1CNC2=C1C=C(C=C2)F

Specifications

Quantity 5 g
Sensitivity Air sensitive
Synonym 5-fluoroindoline, 5-fluoro-2,3-dihydro-1h-indole, 1h-indole,5-fluoro-2,3-dihydro, 1h-indole, 5-fluoro-2,3-dihydro, 5-fluoro-indoline, peakdale1_000398, 5-fluoro-2,3-dihydro-1h-indol, 2,3-dihydro-5-fluoro-1h-indole
Solubility Information Slightly soluble in water.
Formula Weight 137.16
Percent Purity 97%
Chemical Name or Material 5-Fluoroindoline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.