Learn More
5-Fluoroindole-2-carboxylic acid, 98+%, Thermo Scientific Chemicals
35.07 € - 450.00 €
Chemical Identifiers
CAS | 399-76-8 |
---|---|
Molecular Formula | C9H6FNO2 |
Molecular Weight (g/mol) | 179.15 |
MDL Number | MFCD00005612 |
InChI Key | WTXBRZCVLDTWLP-UHFFFAOYSA-N |
Synonym | 5-fluoroindole-2-carboxylic acid, 2-carboxy-5-fluoroindole, 1h-indole-2-carboxylic acid, 5-fluoro, chembl23507, spectrum_001495, pubchem1683, specplus_000678, opera_id_1340, spectrum2_001469, spectrum3_001043 |
PubChem CID | 1820 |
IUPAC Name | 5-fluoro-1H-indole-2-carboxylic acid |
SMILES | C1=CC2=C(C=C1F)C=C(N2)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11415153
|
Thermo Scientific Alfa Aesar
A12794.03 |
1 g |
35.07 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11425153
|
Thermo Scientific Alfa Aesar
A12794.06 |
5 g |
140.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11435153
|
Thermo Scientific Alfa Aesar
A12794.14 |
25 g |
450.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
5-Fluoroindole-2-carboxylic acid,is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Fluoroindole-2-carboxylic acid,is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
It is highly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperatures and pressures.
Chemical Identifiers
399-76-8 | |
179.15 | |
WTXBRZCVLDTWLP-UHFFFAOYSA-N | |
1820 | |
C1=CC2=C(C=C1F)C=C(N2)C(=O)O |
C9H6FNO2 | |
MFCD00005612 | |
5-fluoroindole-2-carboxylic acid, 2-carboxy-5-fluoroindole, 1h-indole-2-carboxylic acid, 5-fluoro, chembl23507, spectrum_001495, pubchem1683, specplus_000678, opera_id_1340, spectrum2_001469, spectrum3_001043 | |
5-fluoro-1H-indole-2-carboxylic acid |
Specifications
399-76-8 | |
C9H6FNO2 | |
153217 | |
It is highly soluble in water. | |
C1=CC2=C(C=C1F)C=C(N2)C(=O)O | |
179.15 | |
179.15 | |
5-Fluoroindole-2-carboxylic acid |
∼259°C (decomposition) | |
MFCD00005612 | |
5-fluoroindole-2-carboxylic acid, 2-carboxy-5-fluoroindole, 1h-indole-2-carboxylic acid, 5-fluoro, chembl23507, spectrum_001495, pubchem1683, specplus_000678, opera_id_1340, spectrum2_001469, spectrum3_001043 | |
WTXBRZCVLDTWLP-UHFFFAOYSA-N | |
5-fluoro-1H-indole-2-carboxylic acid | |
1820 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 206-919-5
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- 5-Fluoroindole-2-carboxylic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : 5-Fluoro-1H-indole-2-carboxylic acid
RUO – Research Use Only