missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Cyano-2-(trifluoromethoxy)benzeneboronic acid, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 1072946-64-5 |
---|---|
Molecular Formula | C8H5BF3NO3 |
Molecular Weight (g/mol) | 230.937 |
InChI Key | SZPKBNMEIASWNB-UHFFFAOYSA-N |
Synonym | 5-cyano-2-trifluoromethoxy phenylboronic acid, 5-cyano-2-trifluoromethoxy phenyl boronic acid, acmc-2098sk |
PubChem CID | 46738976 |
IUPAC Name | [5-cyano-2-(trifluoromethoxy)phenyl]boronic acid |
SMILES | B(C1=C(C=CC(=C1)C#N)OC(F)(F)F)(O)O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1072946-64-5 | |
230.937 | |
5-cyano-2-trifluoromethoxy phenylboronic acid, 5-cyano-2-trifluoromethoxy phenyl boronic acid, acmc-2098sk | |
[5-cyano-2-(trifluoromethoxy)phenyl]boronic acid |
C8H5BF3NO3 | |
SZPKBNMEIASWNB-UHFFFAOYSA-N | |
46738976 | |
B(C1=C(C=CC(=C1)C#N)OC(F)(F)F)(O)O |
Specifications
1072946-64-5 | |
5-cyano-2-trifluoromethoxy phenylboronic acid, 5-cyano-2-trifluoromethoxy phenyl boronic acid, acmc-2098sk | |
B(C1=C(C=CC(=C1)C#N)OC(F)(F)F)(O)O | |
230.937 | |
230.94 | |
Solid |
C8H5BF3NO3 | |
SZPKBNMEIASWNB-UHFFFAOYSA-N | |
[5-cyano-2-(trifluoromethoxy)phenyl]boronic acid | |
46738976 | |
97% | |
5-Cyano-2-(trifluoromethoxy)benzeneboronic acid |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title