missing translation for 'onlineSavingsMsg'
Learn More

5-chlorobenzo[d]isoxazol-3-ol, Thermo Scientific™

72.50 € - 145.00 €

Chemical Identifiers

CAS 24603-63-2
Molecular Formula C7H4ClNO2
Molecular Weight (g/mol) 169.564
InChI Key WIZWHBRFTCYPDN-UHFFFAOYSA-N
Synonym 5-chlorobenzo d isoxazol-3-ol, 5-chloro-3-hydroxybenzisoxazole, 5-chloro-1,2-benzoxazol-3-ol, 5-chloro-1,2-benzisoxazol-3 2h-one, 1,2-benzisoxazol-3 2h-one, 5-chloro, 5-chloro-2h-1,2-benzoxazol-3-one, pubchem24257, 5-chloro-benzo d isoxazol-3-ol, 5-chloro-benzo d isoxazol-3-one, 5-chlorobenzo d isoxazol-3 2h-one
PubChem CID 3714169
IUPAC Name 5-chloro-1,2-benzoxazol-3-one
SMILES C1=CC2=C(C=C1Cl)C(=O)NO2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10361810
View Documents Promotion Details
Thermo Scientific Maybridge
RF01279CB
250 mg
72.50 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10784292
View Documents Promotion Details
Thermo Scientific Maybridge
RF01279DA
1 g
145.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

24603-63-2
169.564
5-chlorobenzo d isoxazol-3-ol, 5-chloro-3-hydroxybenzisoxazole, 5-chloro-1,2-benzoxazol-3-ol, 5-chloro-1,2-benzisoxazol-3 2h-one, 1,2-benzisoxazol-3 2h-one, 5-chloro, 5-chloro-2h-1,2-benzoxazol-3-one, pubchem24257, 5-chloro-benzo d isoxazol-3-ol, 5-chloro-benzo d isoxazol-3-one, 5-chlorobenzo d isoxazol-3 2h-one
5-chloro-1,2-benzoxazol-3-one
C7H4ClNO2
WIZWHBRFTCYPDN-UHFFFAOYSA-N
3714169
C1=CC2=C(C=C1Cl)C(=O)NO2

Specifications

24603-63-2
Amber Glass Bottle
5-chlorobenzo d isoxazol-3-ol, 5-chloro-3-hydroxybenzisoxazole, 5-chloro-1,2-benzoxazol-3-ol, 5-chloro-1,2-benzisoxazol-3 2h-one, 1,2-benzisoxazol-3 2h-one, 5-chloro, 5-chloro-2h-1,2-benzoxazol-3-one, pubchem24257, 5-chloro-benzo d isoxazol-3-ol, 5-chloro-benzo d isoxazol-3-one, 5-chlorobenzo d isoxazol-3 2h-one
C1=CC2=C(C=C1Cl)C(=O)NO2
169.564
169.57
5-Chlorobenzo[d]isoxazol-3-ol
97%
C7H4ClNO2
WIZWHBRFTCYPDN-UHFFFAOYSA-N
5-chloro-1,2-benzoxazol-3-one
3714169
97%
Videos
SDS
Documents

Documents

Product Certifications