Learn More
5-chloro-3-phenyl-2,1-benzisoxazole, 97%, Thermo Scientific™
11.05 € - 25.62 €
Chemical Identifiers
CAS | 719-64-2 |
---|---|
Molecular Formula | C13H8ClNO |
Molecular Weight (g/mol) | 229.67 |
InChI Key | MUHJZJKVEQASGY-UHFFFAOYSA-N |
Synonym | 5-chloro-3-phenylbenzo c isoxazole, 5-chloro-3-phenylanthranil, 5-chloro-3-phenyl-2,1-benzisoxazole, 3-phenyl-5-chloroanthranil, pubchem8713, acmc-209okx, maybridge1_004397, 2, 5-chloro-3-phenyl, 5-chloro-3-phenyl-benzo c isoxazole, 3-phenyl-5-chloro-2,1-benzisoxazole |
PubChem CID | 347291 |
IUPAC Name | 5-chloro-3-phenyl-2,1-benzoxazole |
SMILES | C1=CC=C(C=C1)C2=C3C=C(C=CC3=NO2)Cl |
Description
Chemical Identifiers
719-64-2 | |
229.67 | |
5-chloro-3-phenylbenzo c isoxazole, 5-chloro-3-phenylanthranil, 5-chloro-3-phenyl-2,1-benzisoxazole, 3-phenyl-5-chloroanthranil, pubchem8713, acmc-209okx, maybridge1_004397, 2, 5-chloro-3-phenyl, 5-chloro-3-phenyl-benzo c isoxazole, 3-phenyl-5-chloro-2,1-benzisoxazole | |
5-chloro-3-phenyl-2,1-benzoxazole |
C13H8ClNO | |
MUHJZJKVEQASGY-UHFFFAOYSA-N | |
347291 | |
C1=CC=C(C=C1)C2=C3C=C(C=CC3=NO2)Cl |
Specifications
719-64-2 | |
100.0 | |
Yellow | |
96% min. (GC) | |
C13H8ClNO | |
MUHJZJKVEQASGY-UHFFFAOYSA-N | |
5-chloro-3-phenyl-2,1-benzoxazole | |
347291 | |
97% | |
5-Chloro-3-phenyl-2, 1-benzisoxazole |
96.0 | |
115.3°C to 116.1°C | |
Authentic | |
Glass Bottle | |
5-chloro-3-phenylbenzo c isoxazole, 5-chloro-3-phenylanthranil, 5-chloro-3-phenyl-2,1-benzisoxazole, 3-phenyl-5-chloroanthranil, pubchem8713, acmc-209okx, maybridge1_004397, 2, 5-chloro-3-phenyl, 5-chloro-3-phenyl-benzo c isoxazole, 3-phenyl-5-chloro-2,1-benzisoxazole | |
C1=CC=C(C=C1)C2=C3C=C(C=CC3=NO2)Cl | |
229.67 | |
229.67 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 211-950-2
- 5-Chloro-3-phenyl-2,1-benzisoxazole
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.