missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carbonitrile, 99%, Thermo Scientific™

Product Code. 11400030
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51626-33-6
Molecular Formula C11H8ClN3
Molecular Weight (g/mol) 217.656
MDL Number MFCD00455809
InChI Key MWXQWRGAGVHIDR-UHFFFAOYSA-N
Synonym 5-chloro-3-methyl-1-phenyl-1h-pyrazole-4-carbonitrile, acmc-20am4a
PubChem CID 2748811
IUPAC Name 5-chloro-3-methyl-1-phenylpyrazole-4-carbonitrile
SMILES CC1=NN(C(=C1C#N)Cl)C2=CC=CC=C2

Specifications

Quantity 5 g
UN Number UN3439
Formula Weight 217.66
Percent Purity 99%
Chemical Name or Material 5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carbonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'