missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methylindole, 98%, Thermo Scientific Chemicals

Product Code. 11470127
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1075-35-0
Molecular Formula C9H8ClN
Molecular Weight (g/mol) 165.62
MDL Number MFCD00005619
InChI Key WUVWAXJXPRYUME-UHFFFAOYSA-N
Synonym 5-chloro-2-methylindole, 2-methyl-5-chloroindole, 1h-indole, 5-chloro-2-methyl, 5-chloro-2-methyl-indole, 2-methyl-5-chloro-1h-indole, pubchem17201, acmc-1bpvo, 5-chlor-2-methyl-1h-indole, indole, 5-chloro-2-methyl, 1h-indole,5-chloro-2-methyl
PubChem CID 70636
IUPAC Name 5-chloro-2-methyl-1H-indole
SMILES CC1=CC2=C(N1)C=CC(=C2)Cl

Specifications

Melting Point 110°C to 114°C
Quantity 25 g
Formula Weight 165.62
Percent Purity 98%
Chemical Name or Material 5-Chloro-2-methylindole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'