missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1,3-dimethyl-1H-pyrazole, 98%, Thermo Scientific Chemicals

83.01 € - 310.55 €

Chemical Identifiers

CAS 54454-10-3
Molecular Formula C5H7ClN2
Molecular Weight (g/mol) 130.58
MDL Number MFCD00051650
InChI Key DDUSLFAWARYAPR-UHFFFAOYSA-N
Synonym 5-chloro-1,3-dimethyl-1h-pyrazole, 1h-pyrazole, 5-chloro-1,3-dimethyl, 1,3-dimethyl-5-chloropyrazole, pubchem9980, acmc-20am54, 5-chloro-1,3-dimethyl-pyrazole, 5-chloranyl-1,3-dimethyl-pyrazole, 1,3-dimethyl-5-chloro-1h-pyrazole, 1h-pyrazole,5-chloro-1,3-dimethyl, 5-chloro-1,3-dimethyl-1h-pyrazole #
PubChem CID 521507
IUPAC Name 5-chloro-1,3-dimethylpyrazole
SMILES CN1N=C(C)C=C1Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11459253
SDS View Documents
Thermo Scientific Alfa Aesar
A11730.06
5 g
83.01 €
5g
Please to purchase this item. Need a web account? Register with us today!
11469253
SDS View Documents
Thermo Scientific Alfa Aesar
A11730.14
25 g
310.55 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

54454-10-3
130.58
DDUSLFAWARYAPR-UHFFFAOYSA-N
521507
CN1N=C(C)C=C1Cl
C5H7ClN2
MFCD00051650
5-chloro-1,3-dimethyl-1h-pyrazole, 1h-pyrazole, 5-chloro-1,3-dimethyl, 1,3-dimethyl-5-chloropyrazole, pubchem9980, acmc-20am54, 5-chloro-1,3-dimethyl-pyrazole, 5-chloranyl-1,3-dimethyl-pyrazole, 1,3-dimethyl-5-chloro-1h-pyrazole, 1h-pyrazole,5-chloro-1,3-dimethyl, 5-chloro-1,3-dimethyl-1h-pyrazole #
5-chloro-1,3-dimethylpyrazole

Specifications

54454-10-3
153°C to 155°C
1.486
113451
DDUSLFAWARYAPR-UHFFFAOYSA-N
5-chloro-1,3-dimethylpyrazole
521507
98%
1.137
C5H7ClN2
MFCD00051650
5-chloro-1,3-dimethyl-1h-pyrazole, 1h-pyrazole, 5-chloro-1,3-dimethyl, 1,3-dimethyl-5-chloropyrazole, pubchem9980, acmc-20am54, 5-chloro-1,3-dimethyl-pyrazole, 5-chloranyl-1,3-dimethyl-pyrazole, 1,3-dimethyl-5-chloro-1h-pyrazole, 1h-pyrazole,5-chloro-1,3-dimethyl, 5-chloro-1,3-dimethyl-1h-pyrazole #
CN1N=C(C)C=C1Cl
130.58
130.58
5-Chloro-1,3-dimethyl-1H-pyrazole
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Chloro-1,3-dimethyl-1H-pyrazole, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.