Learn More
5-Bromosalicylaldehyde, 98%, Thermo Scientific Chemicals
33.17 € - 334.67 €
Chemical Identifiers
CAS | 1761-61-1 |
---|---|
Molecular Formula | C7H5BrO2 |
Molecular Weight (g/mol) | 201.019 |
MDL Number | MFCD00003330 |
InChI Key | MKKSTJKBKNCMRV-UHFFFAOYSA-N |
Synonym | 5-bromosalicylaldehyde, benzaldehyde, 5-bromo-2-hydroxy, 2-hydroxy-5-bromobenzaldehyde, salicylaldehyde, 5-bromo, 5-bromo-salicyclaldehyde, unii-0z65a0a8ce, 5-bromo-2-hydroxy-benzaldehyde, 5-bromo-salicyclic aldehyde, 5-bromo-2-hydroxy benzaldehyde, pubchem3779 |
PubChem CID | 72863 |
IUPAC Name | 5-bromo-2-hydroxybenzaldehyde |
SMILES | C1=CC(=C(C=C1Br)C=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11413904
|
Thermo Scientific Alfa Aesar
A11917.14 |
25 g |
33.17 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11423904
|
Thermo Scientific Alfa Aesar
A11917.22 |
100 g |
89.75 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11433904
|
Thermo Scientific Alfa Aesar
A11917.36 |
500 g |
334.67 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1761-61-1 | |
201.019 | |
MKKSTJKBKNCMRV-UHFFFAOYSA-N | |
72863 | |
C1=CC(=C(C=C1Br)C=O)O |
C7H5BrO2 | |
MFCD00003330 | |
5-bromosalicylaldehyde, benzaldehyde, 5-bromo-2-hydroxy, 2-hydroxy-5-bromobenzaldehyde, salicylaldehyde, 5-bromo, 5-bromo-salicyclaldehyde, unii-0z65a0a8ce, 5-bromo-2-hydroxy-benzaldehyde, 5-bromo-salicyclic aldehyde, 5-bromo-2-hydroxy benzaldehyde, pubchem3779 | |
5-bromo-2-hydroxybenzaldehyde |
Specifications
1761-61-1 | |
C7H5BrO2 | |
774710 | |
5-bromosalicylaldehyde, benzaldehyde, 5-bromo-2-hydroxy, 2-hydroxy-5-bromobenzaldehyde, salicylaldehyde, 5-bromo, 5-bromo-salicyclaldehyde, unii-0z65a0a8ce, 5-bromo-2-hydroxy-benzaldehyde, 5-bromo-salicyclic aldehyde, 5-bromo-2-hydroxy benzaldehyde, pubchem3779 | |
C1=CC(=C(C=C1Br)C=O)O | |
201.019 | |
201.02 | |
5-Bromosalicylaldehyde |
103°C to 106°C | |
MFCD00003330 | |
Air Sensitive | |
MKKSTJKBKNCMRV-UHFFFAOYSA-N | |
5-bromo-2-hydroxybenzaldehyde | |
72863 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 217-167-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 5-Bromosalicylaldehyde
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : 5-Bromosalicylaldehyde
RUO – Research Use Only