missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoindole, 99%, Thermo Scientific Chemicals

Product Code. 11419826
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10075-50-0
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.05
MDL Number MFCD00005670
InChI Key VXWVFZFZYXOBTA-UHFFFAOYSA-N
Synonym 5-bromoindole, 1h-indole, 5-bromo, 5-bromo indole, 5-bi, 5bromoindole, 5-bromo-indole, zlchem 227, 5-bromo 1h indole, pubchem1661
PubChem CID 24905
IUPAC Name 5-bromo-1H-indole
SMILES BrC1=CC=C2NC=CC2=C1

Specifications

Melting Point 89°C to 92°C
Odor Odorless
Quantity 25 g
Beilstein 112877
Formula Weight 196.05
Percent Purity 99%
Chemical Name or Material 5-Bromoindole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'