missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoindole-2-carboxylic acid, 98%, Thermo Scientific Chemicals

35.58 € - 362.00 €

Chemical Identifiers

CAS 7254-19-5
Molecular Formula C9H6BrNO2
Molecular Weight (g/mol) 240.05
MDL Number MFCD00022705
InChI Key YAULOOYNCJDPPU-UHFFFAOYSA-N
Synonym 5-bromoindole-2-carboxylic acid, 5-bromo-2-indolecarboxylic acid, 1h-indolecarboxylic acid, 5-bromo, 1h-indole-2-carboxylic acid, 5-bromo, pubchem1663, zlchem 1343, acmc-1bb5x, chembl22435, 5-bromo indole carboxylic acid, 5-bromanyl-1h-indole-2-carboxylic acid
PubChem CID 252137
IUPAC Name 5-bromo-1H-indole-2-carboxylic acid
SMILES C1=CC2=C(C=C1Br)C=C(N2)C(=O)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10654355
SDS View Documents
Thermo Scientific Acros
398652500
250 mg Glass Bottle
35.58 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10244524
SDS View Documents
Thermo Scientific Acros
398650010
1 g Glass bottle N/A N/A N/A
10001214
SDS View Documents
Thermo Scientific Acros
398650050
5 g Glass bottle N/A N/A N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Bromoindole-2-carboxylic acid, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.