missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-7-nitroindoline, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
5-Bromo-7-nitroindoline is used as pharmaceuticals and intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Bromo-7-nitroindoline is used as pharmaceuticals and intermediates.
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
5-Bromo-7-nitroindoline is used as pharmaceuticals and intermediates.
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
Melting Point | 128°C to 133°C |
Quantity | 1 g |
UN Number | UN2811 |
Beilstein | 1373199 |
Solubility Information | Insoluble in water. |
Formula Weight | 243.06 |
Percent Purity | 98% |
Chemical Name or Material | 5-Bromo-7-nitroindoline |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'