missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-7-nitroindoline, 98%, Thermo Scientific Chemicals

Product Code. 11303896
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

5-Bromo-7-nitroindoline is used as pharmaceuticals and intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-7-nitroindoline is used as pharmaceuticals and intermediates.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80166-90-1
Molecular Formula C8H7BrN2O2
Molecular Weight (g/mol) 243.06
MDL Number MFCD00005708
InChI Key VXKXMHDXFLFIFI-UHFFFAOYSA-N
Synonym 5-bromo-7-nitroindoline, 1h-indole, 5-bromo-2,3-dihydro-7-nitro, 5-bromo-7-nitro-1h indoline, 5-bromo-7-nitro-2,3-dihydroindole, 5-bromo-2,3-dihydro-7-nitro-1h-indole, pubchem7507, 5-bromo-7-nitro-indoline
PubChem CID 3018911
IUPAC Name 5-bromo-7-nitro-2,3-dihydro-1H-indole
SMILES [O-][N+](=O)C1=CC(Br)=CC2=C1NCC2

Specifications

Melting Point 128°C to 133°C
Quantity 5 g
UN Number UN2811
Beilstein 1373199
Solubility Information Insoluble in water.
Formula Weight 243.06
Percent Purity 98%
Chemical Name or Material 5-Bromo-7-nitroindoline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'