missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-methoxyquinoline, 96%, Thermo Scientific Chemicals

Product Code. 15401068
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate.

Notes
Refrigerated (2-4°C)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 776296-12-9
Molecular Formula C10H8BrNO
Molecular Weight (g/mol) 238.084
MDL Number MFCD16038652
InChI Key YNVIAXULFXXTLV-UHFFFAOYSA-N
Synonym 5-bromo-3-methoxy-quinoline, 5-bromo-3-methyoxy-quinoline, quinoline, 5-bromo-3-methoxy
PubChem CID 58227851
IUPAC Name 5-bromo-3-methoxyquinoline
SMILES COC1=CN=C2C=CC=C(C2=C1)Br

Specifications

Quantity 5 g
Formula Weight 238.08
Percent Purity 96%
Chemical Name or Material 5-Bromo-3-methoxyquinoline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'