missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-iodopyrimidine, 98%, Thermo Scientific Chemicals

Product Code. 11348426
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 183438-24-6
Molecular Formula C4H2BrIN2
Molecular Weight (g/mol) 284.882
MDL Number MFCD01318111
InChI Key ZEZKXPQIDURFKA-UHFFFAOYSA-N
Synonym 5-bromo-2-iodo-pyrimidine, 2-iodo-5-bromopyrimidine, 5-bromo-2-iodo pyrimidine, pyrimidine, 5-bromo-2-iodo, 5-bromo-2-iodpyrimidine, 5-bromo-2-iodo-1,3-diazine, pubchem6900, 5-brmo-2-iodopyrimidine, acmc-1c1ed, 2-iodo-5-bromo-pyrimidine
PubChem CID 7006651
IUPAC Name 5-bromo-2-iodopyrimidine
SMILES C1=C(C=NC(=N1)I)Br

Specifications

Melting Point 100°C to 102°C
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 284.88
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-iodopyrimidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'