Learn More
5-Bromo-2-fluoropyridine, 98%, Thermo Scientific Chemicals
93.10 € - 338.00 €
Chemical Identifiers
CAS | 766-11-0 |
---|---|
Molecular Formula | C5H3BrFN |
Molecular Weight (g/mol) | 175.988 |
MDL Number | MFCD01863742 |
InChI Key | MYUQKYGWKHTRPG-UHFFFAOYSA-N |
Synonym | 2-fluoro-5-bromopyridine, 3-bromo-6-fluoropyridine, 5-bromo-2-fluoro pyridine, 5-bromo-2-fluoro-pyridine, pyridine, 5-bromo-2-fluoro, 2-fluoro-5-bromo pyridine, pubchem3550, 2-fluro-5-bromo-pyridine, acmc-209p4s, bromo-2-fluoropyridine, 5 |
PubChem CID | 2783168 |
IUPAC Name | 5-bromo-2-fluoropyridine |
SMILES | C1=CC(=NC=C1Br)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11378916
|
Thermo Scientific Alfa Aesar
L19537.06 |
5 g |
93.10 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11388916
|
Thermo Scientific Alfa Aesar
L19537.14 |
25 g |
338.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
766-11-0 | |
175.988 | |
MYUQKYGWKHTRPG-UHFFFAOYSA-N | |
2783168 | |
C1=CC(=NC=C1Br)F |
C5H3BrFN | |
MFCD01863742 | |
2-fluoro-5-bromopyridine, 3-bromo-6-fluoropyridine, 5-bromo-2-fluoro pyridine, 5-bromo-2-fluoro-pyridine, pyridine, 5-bromo-2-fluoro, 2-fluoro-5-bromo pyridine, pubchem3550, 2-fluro-5-bromo-pyridine, acmc-209p4s, bromo-2-fluoropyridine, 5 | |
5-bromo-2-fluoropyridine |
Specifications
766-11-0 | |
162°C to 164°C | |
Characteristic | |
1.5294 | |
UN2810 | |
2-fluoro-5-bromopyridine, 3-bromo-6-fluoropyridine, 5-bromo-2-fluoro pyridine, 5-bromo-2-fluoro-pyridine, pyridine, 5-bromo-2-fluoro, 2-fluoro-5-bromo pyridine, pubchem3550, 2-fluro-5-bromo-pyridine, acmc-209p4s, bromo-2-fluoropyridine, 5 | |
C1=CC(=NC=C1Br)F | |
175.988 | |
175.99 | |
5-Bromo-2-fluoropyridine |
1.72 | |
73°C (163°F) | |
C5H3BrFN | |
MFCD01863742 | |
1363171 | |
MYUQKYGWKHTRPG-UHFFFAOYSA-N | |
5-bromo-2-fluoropyridine | |
2783168 | |
98% |
Safety and Handling
GHS H Statement
H311-H227-H302-H332-H315-H319-H335
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P370+P378q-P501c
H227-H302+H332-H311-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only