missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,3-dihydro-1-benzofuran, 97%, Thermo Scientific™

Product Code. 10415312
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 66826-78-6
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.05
MDL Number MFCD02677716
InChI Key UDWFSJAYXTXMLM-UHFFFAOYSA-N
Synonym 5-bromo-2,3-dihydrobenzofuran, 5-bromo-2,3-dihydrobenzo b furan, 5-bromocoumaran, benzofuran, 5-bromo-2,3-dihydro, 2,3-dihydro-5-bromobenzofuran, buttpark 36\18-16, pubchem7055, 5-bromo-2,3 dihydrobenzofuran, acmc-1b981, 5-bromo-2,3-dihydro-benzofuran
PubChem CID 2776159
IUPAC Name 5-bromo-2,3-dihydro-1-benzofuran
SMILES BrC1=CC2=C(OCC2)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 199.05
Percent Purity 97%
Chemical Name or Material 5-Bromo-2, 3-dihydro-1-benzofuran
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'