missing translation for 'onlineSavingsMsg'
Learn More

(5-Bromo-1-benzofuran-2-yl)methanol, ≥97%, Thermo Scientific™

41.90 € - 491.00 €

Chemical Identifiers

CAS 38220-77-8
Molecular Formula C9H7BrO2
Molecular Weight (g/mol) 227.06
MDL Number MFCD06659073
InChI Key JYYWIDBNICYLBN-UHFFFAOYSA-N
Synonym 5-bromo-1-benzofuran-2-yl methanol, 5-bromobenzofuran-2-yl methanol, 5-bromobenzo d furan-2-yl methan-1-ol, 5-bromo-2-benzofuranylmethanol, 5-bromo-2-benzofuranyl methanol, 5-bromo-benzofuran-2-yl-methanol, 5-bromo-2-hydroxymethyl-1-benzofuran, 5-bromo-1-benzo b furan-2-yl methanol
PubChem CID 2795537
IUPAC Name (5-bromo-1-benzofuran-2-yl)methanol
SMILES OCC1=CC2=CC(Br)=CC=C2O1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10026292
View Documents
Thermo Scientific Maybridge
CC31209CB
250 mg
41.90 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10321143
View Documents
Thermo Scientific Maybridge
CC31209DA
1 g
99.80 €
1g
Please to purchase this item. Need a web account? Register with us today!
10624405
View Documents
Thermo Scientific Maybridge
CC31209DE
5 g
491.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

38220-77-8
227.06
JYYWIDBNICYLBN-UHFFFAOYSA-N
2795537
OCC1=CC2=CC(Br)=CC=C2O1
C9H7BrO2
MFCD06659073
5-bromo-1-benzofuran-2-yl methanol, 5-bromobenzofuran-2-yl methanol, 5-bromobenzo d furan-2-yl methan-1-ol, 5-bromo-2-benzofuranylmethanol, 5-bromo-2-benzofuranyl methanol, 5-bromo-benzofuran-2-yl-methanol, 5-bromo-2-hydroxymethyl-1-benzofuran, 5-bromo-1-benzo b furan-2-yl methanol
(5-bromo-1-benzofuran-2-yl)methanol

Specifications

38220-77-8
100.0
Amber glass bottle
MFCD06659073
JYYWIDBNICYLBN-UHFFFAOYSA-N
(5-bromo-1-benzofuran-2-yl)methanol
2795537
97%
96.5
97%
C9H7BrO2
5-bromo-1-benzofuran-2-yl methanol, 5-bromobenzofuran-2-yl methanol, 5-bromobenzo d furan-2-yl methan-1-ol, 5-bromo-2-benzofuranylmethanol, 5-bromo-2-benzofuranyl methanol, 5-bromo-benzofuran-2-yl-methanol, 5-bromo-2-hydroxymethyl-1-benzofuran, 5-bromo-1-benzo b furan-2-yl methanol
OCC1=CC2=CC(Br)=CC=C2O1
227.06
227.06
(5-Bromo-1-benzofuran-2-yl)methanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications