missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1-methyl-1H-pyrazole-4-carboxamide, Thermo Scientific™

24.20 € - 62.80 €

Chemical Identifiers

CAS 18213-75-7
Molecular Formula C5H8N4O
Molecular Weight (g/mol) 140.146
MDL Number MFCD00111808
InChI Key JGSQVTVXGXOSCH-UHFFFAOYSA-N
PubChem CID 265696
IUPAC Name 5-amino-1-methylpyrazole-4-carboxamide
SMILES CN1C(=C(C=N1)C(=O)N)N
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10763053
View Documents
Thermo Scientific Maybridge
KM00775CB
250 mg Amber Glass Bottle
24.20 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10600283
View Documents
Thermo Scientific Maybridge
KM00775DA
1 g Amber Glass Bottle
62.80 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

18213-75-7
140.146
JGSQVTVXGXOSCH-UHFFFAOYSA-N
5-amino-1-methylpyrazole-4-carboxamide
C5H8N4O
MFCD00111808
265696
CN1C(=C(C=N1)C(=O)N)N

Specifications

18213-75-7
95%
MFCD00111808
JGSQVTVXGXOSCH-UHFFFAOYSA-N
5-amino-1-methylpyrazole-4-carboxamide
265696
95%
95.0
C5H8N4O
5-amino-1-methyl-1h-pyrazole-4-carboxamide, 1h-pyrazole-4-carboxamide, 5-amino-1-methyl, pubchem22392, 5-amino-1-methyl-4-pyrazolecarboxamide, 5-azanyl-1-methyl-pyrazole-4-carboxamide, 1-methyl-5-amino-1h-pyrazole-4-carboxamide, 1h-pyrazole-4-carboxamide,5-amino-1-methyl, 5-imino-1-methyl-2h-pyrazole-4-carboxamide, 3-imino-2-methyl-2,3-dihydro-1h-pyrazole-4-carboxamide
CN1C(=C(C=N1)C(=O)N)N
140.146
140.15
5-Amino-1-methyl-1H-pyrazole-4-carboxamide
Videos
SDS
Documents

Documents

Product Certifications