missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Allyl-2-hydroxy-3-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
81.81 € - 169.85 €
Chemical Identifiers
CAS | 22934-51-6 |
---|---|
Molecular Formula | C11H12O3 |
Molecular Weight (g/mol) | 192.21 |
MDL Number | MFCD06408022 |
InChI Key | FDHXEIOBIOVBEN-UHFFFAOYSA-N |
Synonym | 5-allyl-2-hydroxy-3-methoxybenzaldehyde, 5-allyl-3-methoxysalicylaldehyde, benzaldehyde, 2-hydroxy-3-methoxy-5-2-propenyl, benzaldehyde,2-hydroxy-3-methoxy-5-2-propen-1-yl, 2-hydroxy-3-methoxy-5-prop-2-en-1-yl benzaldehyde, formyl eugenol, acmc-209fzq, 2-hydroxy-3-methoxy-5-2-propenyl benzaldehyde, 3-methoxy-2-oxidanyl-5-prop-2-enyl-benzaldehyde |
PubChem CID | 1713887 |
IUPAC Name | 2-hydroxy-3-methoxy-5-prop-2-enylbenzaldehyde |
SMILES | COC1=C(C(=CC(=C1)CC=C)C=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10797913
|
Thermo Scientific Acros
379360100 |
10 g | Glass bottle |
81.81 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10725233
|
Thermo Scientific Acros
379360250 |
25 g | Glass bottle |
169.85 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
22934-51-6 | |
192.21 | |
FDHXEIOBIOVBEN-UHFFFAOYSA-N | |
1713887 | |
COC1=C(C(=CC(=C1)CC=C)C=O)O |
C11H12O3 | |
MFCD06408022 | |
5-allyl-2-hydroxy-3-methoxybenzaldehyde, 5-allyl-3-methoxysalicylaldehyde, benzaldehyde, 2-hydroxy-3-methoxy-5-2-propenyl, benzaldehyde,2-hydroxy-3-methoxy-5-2-propen-1-yl, 2-hydroxy-3-methoxy-5-prop-2-en-1-yl benzaldehyde, formyl eugenol, acmc-209fzq, 2-hydroxy-3-methoxy-5-2-propenyl benzaldehyde, 3-methoxy-2-oxidanyl-5-prop-2-enyl-benzaldehyde | |
2-hydroxy-3-methoxy-5-prop-2-enylbenzaldehyde |
Specifications
22934-51-6 | |
100.0 | |
Yellow | |
97.5% min. (GC) | |
MFCD06408022 | |
Solubility in water: insoluble | |
COC1=C(C(=CC(=C1)CC=C)C=O)O | |
192.21 | |
192.21 | |
Crystalline Powder |
97.5 | |
48.0°C to 52.0°C | |
Authentic | |
C11H12O3 | |
5-allyl-2-hydroxy-3-methoxybenzaldehyde, 5-allyl-3-methoxysalicylaldehyde, benzaldehyde, 2-hydroxy-3-methoxy-5-2-propenyl, benzaldehyde,2-hydroxy-3-methoxy-5-2-propen-1-yl, 2-hydroxy-3-methoxy-5-prop-2-en-1-yl benzaldehyde, formyl eugenol, acmc-209fzq, 2-hydroxy-3-methoxy-5-2-propenyl benzaldehyde, 3-methoxy-2-oxidanyl-5-prop-2-enyl-benzaldehyde | |
FDHXEIOBIOVBEN-UHFFFAOYSA-N | |
2-hydroxy-3-methoxy-5-prop-2-enylbenzaldehyde | |
1713887 | |
98% | |
5-Allyl-2-hydroxy-3-methoxybenzaldehyde |
Safety and Handling
Product Identifier
- 5-Allyl-2-hydroxy-3-methoxybenzaldehyde
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title