missing translation for 'onlineSavingsMsg'
Learn More

5-Acetyluracil, 98%, Thermo Scientific™

80.40 € - 1342.00 €

Chemical Identifiers

CAS 6214-65-9
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.13
MDL Number MFCD00151968
InChI Key YNYDWEIQSDFDLK-UHFFFAOYSA-N
Synonym 5-acetyluracil, 5-acetylpyrimidine-2,4 1h,3h-dione, 1-2,4-dihydroxypyrimidin-5-yl ethanone, 2,4 1h,3h-pyrimidinedione, 5-acetyl, 5-acetyl-1,3-dihydropyrimidine-2,4-dione, 5-acetyl-1,2,3,4-tetrahydropyrimidine-2,4-dione, 5-acetyl-2,6-dioxo-1,3-dihydropyrimidine, 5-acetyl uracil, acetylpyrimidinedione, zlchem 449
PubChem CID 234581
SMILES CC(=O)C1=CNC(=O)NC1=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11408560
View Documents
Thermo Scientific Alfa Aesar
044378.03
1 g
80.40 €
1g
Please to purchase this item. Need a web account? Register with us today!
11498550
View Documents
Thermo Scientific Alfa Aesar
044378.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11418560
View Documents
Thermo Scientific Alfa Aesar
044378.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

6214-65-9
154.13
YNYDWEIQSDFDLK-UHFFFAOYSA-N
234581
C6H6N2O3
MFCD00151968
5-acetyluracil, 5-acetylpyrimidine-2,4 1h,3h-dione, 1-2,4-dihydroxypyrimidin-5-yl ethanone, 2,4 1h,3h-pyrimidinedione, 5-acetyl, 5-acetyl-1,3-dihydropyrimidine-2,4-dione, 5-acetyl-1,2,3,4-tetrahydropyrimidine-2,4-dione, 5-acetyl-2,6-dioxo-1,3-dihydropyrimidine, 5-acetyl uracil, acetylpyrimidinedione, zlchem 449
CC(=O)C1=CNC(=O)NC1=O

Specifications

6214-65-9
MFCD00151968
YNYDWEIQSDFDLK-UHFFFAOYSA-N
5-acetyl-1H-pyrimidine-2,4-dione
234581
98%
5-Acetyluracil
C6H6N2O3
5-acetyluracil, 5-acetylpyrimidine-2,4 1h,3h-dione, 1-2,4-dihydroxypyrimidin-5-yl ethanone, 2,4 1h,3h-pyrimidinedione, 5-acetyl, 5-acetyl-1,3-dihydropyrimidine-2,4-dione, 5-acetyl-1,2,3,4-tetrahydropyrimidine-2,4-dione, 5-acetyl-2,6-dioxo-1,3-dihydropyrimidine, 5-acetyl uracil, acetylpyrimidinedione, zlchem 449
CC(=O)C1=CNC(=O)NC1=O
154.13
154.11
Powder
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications